2-Fluoro-3-methoxy-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

50657-1G 50657-25G 50657-500MG 50657-5G
76904-594EA 411.63 USD
76904-594 76904-596 76904-598 76904-600
2-Fluoro-3-methoxy-5-(trifluoromethyl)benzoic acid ≥97%
2-Fluoro-3-methoxy-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₆F₄O₃
MW: 238.136 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32902898
CAS Number: 2387276-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR