2-Bromo-3,6-difluorobenzal fluoride ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-3-(difluoromethyl)-1,4-difluorobenzene

50599-1G 50599-25G 50599-500MG 50599-5G
76904-578EA 251.44 USD
76904-578 76904-580 76904-582 76904-584
2-Bromo-3,6-difluorobenzal fluoride ≥97%
2-Bromo-3,6-difluorobenzal fluoride
Formula: C₇H₃BrF₄
MW: 242.996 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28166242
CAS Number: 1672663-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR