2,6-Dichloro-3-methoxybenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

49852-1G 49852-25G 49852-500MG 49852-5G
76904-536EA 155.96 USD
76904-536 76904-538 76904-540 76904-542
2,6-Dichloro-3-methoxybenzoic acid ≥97% research grade
2,6-Dichloro-3-methoxybenzoic acid
Formula: C₈H₆Cl₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD23701500
CAS Number: 32890-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR