2,5-Dibromo-3-methylaniline ≥97%

Supplier: AOBChem USA

Warning

49627-1G 49627-25G 49627-500MG 49627-5G
76904-522EA 429.67 USD
76904-522 76904-524 76904-526 76904-528
2,5-Dibromo-3-methylaniline ≥97%
2,5-Dibromo-3-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇Br₂N
Storage Temperature: Refrigerator
CAS Number: 1806328-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR