1,4-Dibromo-3,5-dichlorobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,5-Dibromo-1,3-dichlorobenzene

49594-1G 49594-25G 49594-500MG 49594-5G
76904-506EA 177.18 USD
76904-506 76904-508 76904-510 76904-512
1,4-Dibromo-3,5-dichlorobenzene ≥97%
1,4-Dibromo-3,5-dichlorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂Cl₂
Storage Temperature: Ambient
CAS Number: 19393-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR