2,6-Dichloro-3-iodo-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

49471-25G 49471-5G
76904-484EA 559.1 USD
76904-484 76904-486
2,6-Dichloro-3-iodo-4-methylpyridine ≥97%
2,6-Dichloro-3-iodo-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₄Cl₂IN
MW: 287.913 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178568
CAS Number: 2643368-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR