2,6-Dibromo-4-fluoro-3-methylaniline ≥97%

Supplier: AOBChem USA

Warning

49203-1G 49203-25G 49203-500MG 49203-5G
76904-436EA 147.47 USD
76904-436 76904-438 76904-440 76904-442
2,6-Dibromo-4-fluoro-3-methylaniline ≥97%
2,6-Dibromo-4-fluoro-3-methylaniline

Precaución: for Research Use Only

Formula: C₇H₆Br₂FN
Storage Temperature: Refrigerator
MDL Number: MFCD09800776
CAS Number: 1000576-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR