2-Fluoro-4-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

49121-1G 49121-25G 49121-5G
76904-422EA 67.9 USD
76904-422 76904-424 76904-426
2-Fluoro-4-methylbenzamide ≥97%
2-Fluoro-4-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD06660328
CAS Number: 886761-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR