2,6-Difluoro-m-toluic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Difluoro-3-methylbenzoic acid

49057-25G 49057-5G
76904-414EA 310.85 USD
76904-414 76904-416
2,6-Difluoro-m-toluic acid ≥97%
2,6-Difluoro-m-toluic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631332
CAS Number: 32890-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR