2-Methoxy-3-(methoxycarbonyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (2-Methoxy-3-(methoxycarbonyl)phenyl)boronic acid

49027-1G 49027-25G 49027-500MG 49027-5G
76904-402EA 292.81 USD
76904-402 76904-404 76904-406 76904-408
2-Methoxy-3-(methoxycarbonyl)phenylboronic acid ≥95%
2-Methoxy-3-(methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₅
MW: 209.992 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23379502
CAS Number: 1332637-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR