2-Iodo-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48950-1G 48950-25G 48950-5G
76904-392EA 84.88 USD
76904-392 76904-394 76904-396
2-Iodo-5-methoxybenzaldehyde ≥97%
2-Iodo-5-methoxybenzaldehyde
Formula: C₈H₇IO₂
MW: 262.046 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18905098
CAS Number: 77287-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR