2-Bromo-3,6-dichlorophenol ≥97%

Supplier: AOBChem USA

Warning

48858-1G 48858-25G 48858-500MG 48858-5G
76904-376EA 292.81 USD
76904-376 76904-378 76904-380 76904-382
2-Bromo-3,6-dichlorophenol ≥97%
2-Bromo-3,6-dichlorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrCl₂O
MW: 241.897 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00152293
CAS Number: 63862-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR