2-Bromo-4-fluoro-5-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

48847-25G 48847-5G
76904-364EA 598.35 USD
76904-364 76904-366
2-Bromo-4-fluoro-5-methoxybenzaldehyde ≥95%
2-Bromo-4-fluoro-5-methoxybenzaldehyde
Formula: C₈H₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD22371190
CAS Number: 865186-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR