2-Bromo-1-(4-fluorophenoxy)-4-nitrobenzene ≥95%

Supplier: AOBChem USA

48731-1G 48731-25G 48731-500MG 48731-5G
76904-350EA 147.47 USD
76904-350 76904-352 76904-354 76904-356
2-Bromo-1-(4-fluorophenoxy)-4-nitrobenzene ≥95%
2-Bromo-1-(4-fluorophenoxy)-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₁₂H₇BrFNO₃
MDL Number: MFCD02081973
CAS Number: 477856-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR