2-Fluoro-6-iodo-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

48561-1G 48561-25G 48561-500MG 48561-5G
76904-334EA 289.63 USD
76904-334 76904-336 76904-338 76904-340
2-Fluoro-6-iodo-3-methylbenzoic acid ≥97%
2-Fluoro-6-iodo-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆FIO₂
MW: 280.035 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11847125
CAS Number: 796967-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR