2-Chloro-3-methoxy-5-methylpyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
48406-1G
48406-500MG
48406-5G
48406-25G
76904-304EA
464.68
USD
76904-304
76904-306
76904-308
77656-164
2-Chloro-3-methoxy-5-methylpyridine ≥97%
2-Chloro-3-methoxy-5-methylpyridine
Formula:
C₇H₈ClNO Storage Temperature: Ambient |
MDL Number:
MFCD13563046 CAS Number: 1203499-46-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...