2-Methoxy-4-(methoxymethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

48339-1G 48339-25G 48339-500MG 48339-5G
76904-256EA 142.17 USD
76904-256 76904-258 76904-260 76904-262
2-Methoxy-4-(methoxymethoxy)benzaldehyde ≥97%
2-Methoxy-4-(methoxymethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06661070
CAS Number: 114628-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR