2,6-Dichloro-4-iodobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

48290-1G 48290-25G 48290-5G
76904-242EA 109.28 USD
76904-242 76904-244 76904-246
2,6-Dichloro-4-iodobenzoic acid ≥95%
2,6-Dichloro-4-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Cl₂IO₂
MW: 316.908 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22690836
CAS Number: 1258298-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR