2-Bromo-1,5-dichloro-3-nitrobenzene ≥95%

Supplier: AOBChem USA

Warning

47774-1G 47774-25G 47774-500MG 47774-5G
76904-202EA 258.86 USD
76904-202 76904-204 76904-206 76904-208
2-Bromo-1,5-dichloro-3-nitrobenzene ≥95%
2-Bromo-1,5-dichloro-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂NO₂
MW: 270.896 g/mol
Storage Temperature: Ambient
CAS Number: 1185916-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR