3-Chloro-2,4-difluoroiodobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-1,3-difluoro-4-iodobenzene

47708-25G 47708-5G
76904-192EA 418 USD
76904-192 76904-194
3-Chloro-2,4-difluoroiodobenzene ≥97%
3-Chloro-2,4-difluoroiodobenzene
Formula: C₆H₂ClF₂I
MW: 274.434 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00142890
CAS Number: 202925-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR