2,6-Dichloro-4-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

47424-1G 47424-25G 47424-500MG 47424-5G
76904-158EA 142.17 USD
76904-158 76904-160 76904-162 76904-164
2,6-Dichloro-4-(methylthio)pyridine ≥97%
2,6-Dichloro-4-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₆H₅Cl₂NS
MW: 194.082 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32707272
CAS Number: 153564-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR