2-n-Propoxypyridine-3-carboxaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Propoxynicotinaldehyde

47367-1G 47367-25G 47367-500MG 47367-5G
76904-144EA 147.47 USD
76904-144 76904-146 76904-148 76904-150
2-n-Propoxypyridine-3-carboxaldehyde ≥95%
2-n-Propoxypyridine-3-carboxaldehyde
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06804561
CAS Number: 885278-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR