2-Ethoxy-6-fluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

46691-1G 46691-25G 46691-500MG 46691-5G
76904-106EA 215.37 USD
76904-106 76904-108 76904-110 76904-112
2-Ethoxy-6-fluorobenzaldehyde ≥95%
2-Ethoxy-6-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD02114885
CAS Number: 82129-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR