2-Chloro-3,5-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

46646-1G 46646-500MG 46646-5G
76904-092EA 467.86 USD
76904-092 76904-094 76904-096
2-Chloro-3,5-difluorobenzaldehyde ≥97%
2-Chloro-3,5-difluorobenzaldehyde
Formula: C₇H₃ClF₂O
MW: 176.548 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18415552
CAS Number: 1261561-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR