2-Chloro-4-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

46373-1G 46373-25G 46373-500MG 46373-5G
76904-076EA 233.4 USD
76904-076 76904-078 76904-080 76904-082
2-Chloro-4-iodobenzaldehyde ≥97%
2-Chloro-4-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClIO
MW: 266.464 g/mol
Storage Temperature: Ambient
CAS Number: 1260810-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR