2-Bromo-3-fluoro-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

46225-25G 46225-5G
76904-046EA 233.4 USD
76904-046 76904-048
2-Bromo-3-fluoro-4-methylpyridine ≥97%
2-Bromo-3-fluoro-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFN
Storage Temperature: Refrigerator
MDL Number: MFCD04112499
CAS Number: 884494-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR