2-Chlorobiphenyl (PCB No. 1) ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-1,1'-biphenyl, PCB No. 1, 2-Monochlorobiphenyl, 2-PCB

45974-25G 45974-5G
76904-028EA 187.78 USD
76904-028 76904-030
2-Chlorobiphenyl (PCB No. 1) ≥97%
2-Chlorobiphenyl (PCB No. 1)

Precaución: for Research Use Only

Formula: C₁₂H₉Cl
MW: 188.66 g/mol
Storage Temperature: Ambient
CAS Number: 2051-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR