2-Chloro-4-fluoro-6-(trifluoromethyl)aniline ≥97%

Supplier: AOBChem USA

Warning

45449-1G 45449-25G 45449-500MG 45449-5G
76903-970EA 381.93 USD
76903-970 76903-972 76903-974 76903-976
2-Chloro-4-fluoro-6-(trifluoromethyl)aniline ≥97%
2-Chloro-4-fluoro-6-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄ClF₄N
MW: 213.56 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16040046
CAS Number: 1235439-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR