2-Chloro-4,5-dimethoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

45153-1G 45153-25G 45153-500MG 45153-5G
76903-954EA 203.7 USD
76903-954 76903-956 76903-958 76903-960
2-Chloro-4,5-dimethoxyphenylboronic acid ≥97%
2-Chloro-4,5-dimethoxyphenylboronic acid
Formula: C₈H₁₀BClO₄
MW: 216.43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23379550
CAS Number: 950662-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR