2,6-Difluoro-3-methylphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

41993-1G 41993-25G 41993-500MG 41993-5G
76903-876EA 159.14 USD
76903-876 76903-878 76903-880 76903-882
2,6-Difluoro-3-methylphenylboronic acid ≥97% research grade
2,6-Difluoro-3-methylphenylboronic acid
Formula: C₇H₇BF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09265479
CAS Number: 1586045-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR