2-Fluoro-6-phenyl-3-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

41797-1G 41797-25G 41797-500MG 41797-5G
76903-860EA 379.81 USD
76903-860 76903-862 76903-864 76903-866
2-Fluoro-6-phenyl-3-pyridineboronic acid ≥97%
2-Fluoro-6-phenyl-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₉BFNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD07437944
CAS Number: 1029654-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR