2-Cyano-6-methoxyphenylboronic acid 97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Cyano-6-methoxyphenylboronic Acid

41184-1G 41184-25G 41184-5G 41184-500MG
76903-846EA 94.43 USD
76903-846 76903-848 76903-850 77650-598
2-Cyano-6-methoxyphenylboronic acid 97%
2-Cyano-6-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₃
MW: 176.965 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10696677
CAS Number: 1164100-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR