2,5-Dichloro-3-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,5-Dichloropyridine-3-boronic acid pinacol ester, 2,5-Dichloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

39059-1G 39059-25G 39059-500MG 39059-5G
76903-754EA 124.13 USD
76903-754 76903-756 76903-758 76903-760
2,5-Dichloro-3-pyridineboronic acid pinacol ester ≥97%
2,5-Dichloro-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₄BCl₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD08063094
CAS Number: 1073371-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR