2,6-Dichloro-4-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

38756-25G 38756-5G
76903-730EA 219.61 USD
76903-730 76903-732
2,6-Dichloro-4-fluorophenol ≥97%
2,6-Dichloro-4-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₃Cl₂FO
MW: 180.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010675
CAS Number: 392-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR