2-Fluoro-6-methoxy-3-methylphenol ≥97%

Supplier: AOBChem USA

Warning

38658-1G 38658-25G 38658-500MG 38658-5G
76903-704EA 296 USD
76903-704 76903-706 76903-708 76903-710
2-Fluoro-6-methoxy-3-methylphenol ≥97%
2-Fluoro-6-methoxy-3-methylphenol

Precaución: for Research Use Only

Formula: C₈H₉FO₂
MW: 156.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28515793
CAS Number: 1782490-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR