2-Fluoro-6-hydroxy-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

38645-1G 38645-500MG 38645-5G
76903-690EA 647.15 USD
76903-690 76903-692 76903-694
2-Fluoro-6-hydroxy-3-methylbenzoic acid ≥97%
2-Fluoro-6-hydroxy-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MW: 170.138 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28519079
CAS Number: 133788-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR