2-Chloro-3-iodo-5-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

38543-1G 38543-25G 38543-500MG 38543-5G
76903-674EA 373.44 USD
76903-674 76903-676 76903-678 76903-680
2-Chloro-3-iodo-5-methylpyridine ≥97%
2-Chloro-3-iodo-5-methylpyridine
Formula: C₆H₅ClIN
Storage Temperature: Ambient
MDL Number: MFCD09839273
CAS Number: 59782-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR