2,6-Dibromo-3-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

38198-1G 38198-25G 38198-5G
76903-602EA 82.76 USD
76903-602 76903-604 76903-606
2,6-Dibromo-3-methoxybenzoic acid ≥97%
2,6-Dibromo-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD32632318
CAS Number: 2091763-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR