2-Bromo-4,5-dichlorophenol ≥97%

Supplier: AOBChem USA

Warning

38090-1G 38090-25G 38090-500MG 38090-5G
76903-590EA 128.37 USD
76903-590 76903-592 76903-594 76903-596
2-Bromo-4,5-dichlorophenol ≥97%
2-Bromo-4,5-dichlorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrCl₂O
Storage Temperature: Refrigerator
MDL Number: MFCD27947800
CAS Number: 2316-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR