2-Ethoxy-3,4-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37908-1G 37908-25G 37908-500MG 37908-5G
76903-576EA 439.22 USD
76903-576 76903-578 76903-580 76903-582
2-Ethoxy-3,4-difluorobenzaldehyde ≥97%
2-Ethoxy-3,4-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28626415
CAS Number: 1552707-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR