2-Bromo-3-chloro-4-fluoro-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

37697-1G 37697-25G 37697-500MG 37697-5G
76903-560EA 272.66 USD
76903-560 76903-562 76903-564 76903-566
2-Bromo-3-chloro-4-fluoro-1-methoxybenzene ≥97%
2-Bromo-3-chloro-4-fluoro-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29044053
CAS Number: 1881321-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR