2,6-Dimethyl-4-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37675-1G 37675-25G 37675-500MG 37675-5G
76903-544EA 220.67 USD
76903-544 76903-546 76903-548 76903-550
2,6-Dimethyl-4-propoxybenzoic acid ≥97%
2,6-Dimethyl-4-propoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
Storage Temperature: Refrigerator
MDL Number: MFCD18426783
CAS Number: 100256-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR