2-Fluoro-5-methylnicotinamide ≥97%

Supplier: AOBChem USA

Warning

37526-1G 37526-25G 37526-500MG 37526-5G
76903-510EA 350.1 USD
76903-510 76903-512 76903-514 76903-516
2-Fluoro-5-methylnicotinamide ≥97%
2-Fluoro-5-methylnicotinamide

Precaución: for Research Use Only

Formula: C₇H₇FN₂O
MW: 154.142 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18907016
CAS Number: 1379014-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR