2-Bromo-6-ethylphenol ≥97%

Supplier: AOBChem USA

Warning

37318-25G 37318-5G
76903-498EA 360.71 USD
76903-498 76903-500
2-Bromo-6-ethylphenol ≥97%
2-Bromo-6-ethylphenol

Precaución: for Research Use Only

Formula: C₈H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD01707689
CAS Number: 24885-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR