2-Fluoro-4,5-dimethoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

37307-1G 37307-25G 37307-5G
76903-484EA 114.58 USD
76903-484 76903-486 76903-488
2-Fluoro-4,5-dimethoxybenzoic acid ≥95%
2-Fluoro-4,5-dimethoxybenzoic acid
Formula: C₉H₉FO₄
MW: 200.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06208568
CAS Number: 79474-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR