3-(3-Bromo-5-methylphenyl)-2-propenoic acid ≥97%

Supplier: AOBChem USA

Warning

37242-1G 37242-25G 37242-500MG 37242-5G
76903-470EA 148.53 USD
76903-470 76903-472 76903-474 76903-476
3-(3-Bromo-5-methylphenyl)-2-propenoic acid ≥97%
3-(3-Bromo-5-methylphenyl)-2-propenoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₂
MW: 241.081 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18392023
CAS Number: 1262018-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR