2-Chloro-N-cyclohexyl-6-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

37205-1G 37205-25G 37205-500MG 37205-5G
76903-440EA 140.04 USD
76903-440 76903-442 76903-444 76903-446
2-Chloro-N-cyclohexyl-6-(trifluoromethyl)benzamide ≥97%
2-Chloro-N-cyclohexyl-6-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₅ClF₃NO
MW: 305.723 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32632162
CAS Number: 2379322-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR