2,8-Dimethylimidazo[1,2-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

37139-1G 37139-25G 37139-500MG 37139-5G
76903-426EA 254.62 USD
76903-426 76903-428 76903-430 76903-432
2,8-Dimethylimidazo[1,2-a]pyridine ≥97%
2,8-Dimethylimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₉H₁₀N₂
MW: 146.189 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11103528
CAS Number: 67625-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR