2-Methylimidazo[1,2-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

37135-25G 37135-5G
76903-414EA 630.18 USD
76903-414 76903-416
2-Methylimidazo[1,2-a]pyridine ≥97%
2-Methylimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02251330
CAS Number: 934-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR