2'-Methylbiphenyl-4-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2'-Methyl-[1,1'-biphenyl]-4-carboxylic acid

37096-25G 37096-5G
76903-402EA 453.01 USD
76903-402 76903-404
2'-Methylbiphenyl-4-carboxylic acid ≥95%
2'-Methylbiphenyl-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Storage Temperature: Refrigerator
CAS Number: 5748-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR